Merck
Chaperone-Independent Peripheral Quality Control of CFTR by RFFL E3 Ligase.
Developmental cell (2018-03-06)
Tsukasa Okiyoneda, Guido Veit, Ryohei Sakai, Misaki Aki, Takeshi Fujihara, Momoko Higashi, Seiko Susuki-Miyata, Masanori Miyata, Norihito Fukuda, Akihiko Yoshida, Haijin Xu, Pirjo M Apaja, Gergely L Lukacs
ZUSAMMENFASSUNG
The peripheral protein quality control (QC) system removes non-native membrane proteins, including ΔF508-CFTR, the most common CFTR mutant in cystic fibrosis (CF), from the plasma membrane (PM) for lysosomal degradation by ubiquitination. It remains unclear how unfolded membrane proteins are recognized and targeted for ubiquitination and how they are removed from the apical PM. Using comprehensive siRNA screens, we identified RFFL, an E3 ubiquitin (Ub) ligase that directly and selectively recognizes unfolded ΔF508-CFTR through its disordered regions. RFFL retrieves the unfolded CFTR from the PM for lysosomal degradation by chaperone-independent K63-linked poly-ubiquitination. RFFL ablation enhanced the functional expression of cell-surface ΔF508-CFTR in the presence of folding corrector molecules, and this effect was further improved by inhibiting the Hsc70-dependent ubiquitination machinery. We propose that multiple peripheral QC mechanisms evolved to dispose of non-native PM proteins and to preserve cellular proteostasis, even at the cost of eliminating partially functional polypeptides.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Anti-Ubiquitin-Antikörper, Lys63-spezifisch, Klon Apu3, monoklonaler Kaninchen-Antikörper, clone Apu3, from rabbit
Sigma-Aldrich
DUB-Inhibitor V, PR-619, Calbiochem, The DUB Inhibitor V, PR-619 controls the biological activity of DUB. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Anti-CFTR-Antikörper, A.S. 1370-1380, Klon M3A7, clone M3A7, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cystic Fibrosis Transmembrane Conductance Regulator Antibody, a.a. 386-412, clone L12B4, clone L12B4, Chemicon®, from mouse
Sigma-Aldrich
Anti-RFFL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution