Merck
  • Role of innate lymphoid cells and dendritic cells in intradermal immunization of the enterovirus antigen.
Role of innate lymphoid cells and dendritic cells in intradermal immunization of the enterovirus antigen.
NPJ vaccines (2019-04-03)
Shengtao Fan, Yun Liao, Yaru Lian, Guorun Jiang, Li Jiang, Chenhong Dong, Erxia Yang, Lichun Wang, Xingli Xu, Min Feng, Ying Zhang, Qihan Li
ZUSAMMENFASSUNG
Enterovirus type 71 (EV71) and coxsackievirus A 16 (CA16) are the major pathogens of human hand, foot, and mouth disease (HFMD). In our previous study, intramuscular immunization with the inactivated EV71 vaccine elicited effective immunity, while immunization with the inactivated CA16 vaccine did not. In this report, we focused on innate immune responses elicited by inactivated EV71 and CA16 antigens administered intradermally or intramuscularly. The distributions of the EV71 and CA16 antigens administered intradermally or intramuscularly were not obviously different, but the antigens were detected for a shorter period of time when administered intradermally. The expression levels of NF-κB pathway signaling molecules, which were identified as being capable of activating DCs, ILCs, and T cells, were higher in the intradermal group than in the intramuscular group. Antibodies for the EV71 and CA16 antigens colocalized with ILCs and DCs in skin and muscle tissues under fluorescence microscopy. Interestingly, ILC colocalization decreased over time, while DC colocalization increased over time. ELISpot analysis showed that coordination between DCs and ILCs contributed to successful adaptive immunity against vaccine antigens in the skin. EV71 and/or CA16 antigen immunization via the intradermal route was more capable of significantly increasing neutralizing antibody titers and activating specific T cell responses than immunization via the intramuscular route. Furthermore, neonatal mice born to mothers immunized with the EV71 and CA16 antigens were 100% protected against wild-type EV71 or CA16 viral challenge. Together, our results provide new insights into the development of vaccines for HFMD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Enterovirus-71-Antikörper (Kreuzreaktivität mit Coxsackie-Virus A16, Klon 422-8D-4C-4D), ascites fluid, clone 422-8D-4C-4D, Chemicon®