Merck
  • Oligodendrocytes regulate presynaptic properties and neurotransmission through BDNF signaling in the mouse brainstem.
Oligodendrocytes regulate presynaptic properties and neurotransmission through BDNF signaling in the mouse brainstem.
eLife (2019-04-19)
Miae Jang, Elizabeth Gould, Jie Xu, Eun Jung Kim, Jun Hee Kim
ZUSAMMENFASSUNG
Neuron-glia communication contributes to the fine-tuning of synaptic functions. Oligodendrocytes near synapses detect and respond to neuronal activity, but their role in synapse development and plasticity remains largely unexplored. We show that oligodendrocytes modulate neurotransmitter release at presynaptic terminals through secretion of brain-derived neurotrophic factor (BDNF). Oligodendrocyte-derived BDNF functions via presynaptic tropomyosin receptor kinase B (TrkB) to ensure fast, reliable neurotransmitter release and auditory transmission in the developing brain. In auditory brainstem slices from Bdnf+/- mice, reduction in endogenous BDNF significantly decreased vesicular glutamate release by reducing the readily releasable pool of glutamate vesicles, without altering presynaptic Ca2+ channel activation or release probability. Using conditional knockout mice, cell-specific ablation of BDNF in oligodendrocytes largely recapitulated this effect, which was recovered by BDNF or TrkB agonist application. This study highlights a novel function for oligodendrocytes in synaptic transmission and their potential role in the activity-dependent refinement of presynaptic properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-vesikulärer Glutamattransporter 1-Antikörper, serum, Chemicon®
Sigma-Aldrich
Anti-MAP2-Antikörper, Klon AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Strychnine hydrochloride
Sigma-Aldrich
Anti-Olig1-Antikörper, ascites fluid, Chemicon®