Merck
  • dBRWD3 Regulates Tissue Overgrowth and Ectopic Gene Expression Caused by Polycomb Group Mutations.
dBRWD3 Regulates Tissue Overgrowth and Ectopic Gene Expression Caused by Polycomb Group Mutations.
PLoS genetics (2016-09-03)
Hsueh-Tzu Shih, Wei-Yu Chen, Kwei-Yan Liu, Zong-Siou Shih, Yi-Jyun Chen, Paul-Chen Hsieh, Kuan-Lin Kuo, Kuo-How Huang, Pang-Hung Hsu, Ya-Wen Liu, Shih-Peng Chan, Hsiu-Hsiang Lee, Yu-Chen Tsai, June-Tai Wu
ZUSAMMENFASSUNG
To maintain a particular cell fate, a unique set of genes should be expressed while another set is repressed. One way to repress gene expression is through Polycomb group (PcG) proteins that compact chromatin into a silent configuration. In addition to cell fate maintenance, PcG proteins also maintain normal cell physiology, for example cell cycle. In the absence of PcG, ectopic activation of the PcG-repressed genes leads to developmental defects and malignant tumors. Little is known about the molecular nature of ectopic gene expression; especially what differentiates expression of a given gene in the orthotopic tissue (orthotopic expression) and the ectopic expression of the same gene due to PcG mutations. Here we present that ectopic gene expression in PcG mutant cells specifically requires dBRWD3, a negative regulator of HIRA/Yemanuclein (YEM)-mediated histone variant H3.3 deposition. dBRWD3 mutations suppress both the ectopic gene expression and aberrant tissue overgrowth in PcG mutants through a YEM-dependent mechanism. Our findings identified dBRWD3 as a critical regulator that is uniquely required for ectopic gene expression and aberrant tissue overgrowth caused by PcG mutations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Ubiquityl-Histon H2A-Antikörper, Klon E6C5, clone E6C5, Upstate®, from mouse