Merck
  • Activation of the enhancer of zeste homologue 2 gene by the human papillomavirus E7 oncoprotein.
Activation of the enhancer of zeste homologue 2 gene by the human papillomavirus E7 oncoprotein.
Cancer research (2008-12-03)
Daniela Holland, Karin Hoppe-Seyler, Bettina Schuller, Claudia Lohrey, Julia Maroldt, Matthias Dürst, Felix Hoppe-Seyler
ZUSAMMENFASSUNG
The malignant phenotype of human papillomavirus (HPV)-positive cancer cells is maintained by the activity of the viral E6 and E7 genes. Here, we identified the polycomb group gene enhancer of zeste homologue 2 (EZH2) as a novel downstream target for the viral oncogenes in HPV-transformed cells. EZH2 expression was activated by HPV16 E7 at the transcriptional level via E7-mediated release of E2F from pocket proteins. RNA interference analyses showed that continuous EZH2 expression is required for the proliferation of HPV-positive tumor cells by stimulating cell cycle progression at the G1-S boundary. In addition to its growth-promoting activity, EZH2 also contributed to the apoptotic resistance of cervical cancer cells. Furthermore, we found that HPV-positive dysplastic and tumorigenic cervical lesions were characterized by high levels of EZH2 protein in vivo. We conclude that the E7 target gene EZH2 is a major determinant for the proliferation of HPV-positive cancer cells and contributes to their apoptotic resistance. Moreover, EZH2 may serve as a novel therapeutic target for the treatment of cervical cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®