Merck
  • High Glucose Triggers Nucleotide Imbalance through O-GlcNAcylation of Key Enzymes and Induces KRAS Mutation in Pancreatic Cells.
High Glucose Triggers Nucleotide Imbalance through O-GlcNAcylation of Key Enzymes and Induces KRAS Mutation in Pancreatic Cells.
Cell metabolism (2019-03-12)
Chun-Mei Hu, Sui-Chih Tien, Ping-Kun Hsieh, Yung-Ming Jeng, Ming-Chu Chang, Yu-Ting Chang, Yi-Ju Chen, Yu-Ju Chen, Eva Y-H P Lee, Wen-Hwa Lee
ZUSAMMENFASSUNG
KRAS mutations are the earliest events found in approximately 90% of pancreatic ductal adenocarcinomas (PDACs). However, little is known as to why KRAS mutations preferentially occur in PDACs and what processes/factors generate these mutations. While abnormal carbohydrate metabolism is associated with a high risk of pancreatic cancer, it remains elusive whether a direct relationship between KRAS mutations and sugar metabolism exists. Here, we show that under high-glucose conditions, cellular O-GlcNAcylation is significantly elevated in pancreatic cells that exhibit lower phosphofructokinase (PFK) activity than other cell types. This post-translational modification specifically compromises the ribonucleotide reductase (RNR) activity, leading to deficiency in dNTP pools, genomic DNA alterations with KRAS mutations, and cellular transformation. These results establish a mechanistic link between a perturbed sugar metabolism and genomic instability that induces de novo oncogenic KRAS mutations preferentially in pancreatic cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, clone JBW301, Upstate®, from mouse
Roche
Zellproliferationskit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
Anti-53BP1-Antikörper, Klon BP13, clone BP13, Chemicon®, from mouse
Sigma-Aldrich
2′-Deoxycytidine 5′-diphosphate sodium salt, ≥96%
Sigma-Aldrich
Anti-O-GlcNAc Transferase (DM-17) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution