Merck
Whole blueberry protects pancreatic beta-cells in diet-induced obese mouse.
Nutrition & metabolism (2019-05-30)
Weixiang Liu, Yiping Mao, Jacob Schoenborn, Zhihong Wang, Guiliang Tang, Xiaoqing Tang
ZUSAMMENFASSUNG
Blueberry is rich in bioactive substances and possesses powerful antioxidant potential, which can protect against oxidant-induced and inflammatory cell damage and cytotoxicity. The aim of this study was to determine how blueberry affects glucose metabolism and pancreatic β-cell proliferation in high fat diet (HFD)-induced obese mice. Wild type male mice at age of 4 weeks received two different kinds of diets: high-fat diet (HFD) containing 60% fat or modified HFD supplemented with 4% (wt:wt) freeze-dried whole blueberry powder (HFD + B) for 14 weeks. A separate experiment was performed in mice fed with low-fat diet (LFD) containing 10% fat or modified LFD + B supplemented with 4% (wt:wt) freeze-dried whole blueberry powder. The metabolic parameters including blood glucose and insulin levels, glucose and insulin tolerances were measured. Blueberry-supplemented diet significantly increased insulin sensitivity and glucose tolerance in HFD + B mice compared to HFD mice. However, no difference was observed in blood glucose and insulin sensitivity between LFD + B and LFD mice. In addition, blueberry increased β-cell survival and prevented HFD-induced β-cell expansion. The most important finding was the observation of presence of small scattered islets in blueberry treated obese mice, which may reflect a potential role of blueberry in regenerating pancreatic β-cells. Blueberry-supplemented diet can prevent obesity-induced insulin resistance by improving insulin sensitivity and protecting pancreatic β-cells. Blueberry supplementation has the potential to protect and improve health conditions for both type 1 and type 2 diabetes patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
Zellproliferation ELISA, BrdU (chemilumineszierend)
Sigma-Aldrich
Monoklonaler Anti-Insulin-Antikörper in Maus hergestellte Antikörper, clone K36AC10, ascites fluid