Merck
  • TurboID-based proximity labeling reveals that UBR7 is a regulator of N NLR immune receptor-mediated immunity.
TurboID-based proximity labeling reveals that UBR7 is a regulator of N NLR immune receptor-mediated immunity.
Nature communications (2019-07-22)
Yongliang Zhang, Gaoyuan Song, Neeraj K Lal, Ugrappa Nagalakshmi, Yuanyuan Li, Wenjie Zheng, Pin-Jui Huang, Tess C Branon, Alice Y Ting, Justin W Walley, Savithramma P Dinesh-Kumar
ZUSAMMENFASSUNG
Nucleotide-binding leucine-rich repeat (NLR) immune receptors play a critical role in defence against pathogens in plants and animals. However, we know very little about NLR-interacting proteins and the mechanisms that regulate NLR levels. Here, we used proximity labeling (PL) to identify the proteome proximal to N, which is an NLR that confers resistance to Tobacco mosaic virus (TMV). Evaluation of different PL methods indicated that TurboID-based PL provides more efficient levels of biotinylation than BioID and BioID2 in plants. TurboID-based PL of N followed by quantitative proteomic analysis and genetic screening revealed multiple regulators of N-mediated immunity. Interestingly, a putative E3 ubiquitin ligase, UBR7, directly interacts with the TIR domain of N. UBR7 downregulation leads to an increased amount of N protein and enhanced TMV resistance. TMV-p50 effector disrupts the N-UBR7 interaction and relieves negative regulation of N. These findings demonstrate the utility of TurboID-based PL in plants and the N-interacting proteins we identified enhance our understanding of the mechanisms underlying NLR regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Roche
Anti-HA-Peroxidase mit hoher Affinität, from rat IgG1
Millipore
Anti-c-Myc-Agarose-Affinitätsgel in Kaninchen hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Klon 4A6, HRP-Konjugat, clone 4A6, Upstate®, from mouse