Merck
  • Eeyarestatin Compounds Selectively Enhance Sec61-Mediated Ca2+ Leakage from the Endoplasmic Reticulum.
Eeyarestatin Compounds Selectively Enhance Sec61-Mediated Ca2+ Leakage from the Endoplasmic Reticulum.
Cell chemical biology (2019-02-26)
Igor Gamayun, Sarah O'Keefe, Tillman Pick, Marie-Christine Klein, Duy Nguyen, Craig McKibbin, Michela Piacenti, Helen M Williams, Sabine L Flitsch, Roger C Whitehead, Eileithyia Swanton, Volkhard Helms, Stephen High, Richard Zimmermann, Adolfo Cavalié
ZUSAMMENFASSUNG
Eeyarestatin 1 (ES1) inhibits p97-dependent protein degradation, Sec61-dependent protein translocation into the endoplasmic reticulum (ER), and vesicular transport within the endomembrane system. Here, we show that ES1 impairs Ca2+ homeostasis by enhancing the Ca2+ leakage from mammalian ER. A comparison of various ES1 analogs suggested that the 5-nitrofuran (5-NF) ring of ES1 is crucial for this effect. Accordingly, the analog ES24, which conserves the 5-NF domain of ES1, selectively inhibited protein translocation into the ER, displayed the highest potency on ER Ca2+ leakage of ES1 analogs studied and induced Ca2+-dependent cell death. Using small interfering RNA-mediated knockdown of Sec61α, we identified Sec61 complexes as the targets that mediate the gain of Ca2+ leakage induced by ES1 and ES24. By interacting with the lateral gate of Sec61α, ES1 and ES24 likely capture Sec61 complexes in a Ca2+-permeable, open state, in which Sec61 complexes allow Ca2+ leakage but are translocation incompetent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
G 418 -disulfat (Salz) -Lösung, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
ES24, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-SERCa2 ATPase antibody produced in mouse, clone IID8