Merck
  • Corticosteroid-Induced Cutaneous Atrophy and Telangiectasia: Experimental Production Associated With Weight Loss in Rats
Corticosteroid-Induced Cutaneous Atrophy and Telangiectasia: Experimental Production Associated With Weight Loss in Rats
Archives of Dermatological Research (1976)
. Graham Smith, Jr, Richard F. Wehr, et al.