Merck
  • Site-specific phosphorylation of casein kinase 1 δ (CK1δ) regulates its activity towards the circadian regulator PER2.
Site-specific phosphorylation of casein kinase 1 δ (CK1δ) regulates its activity towards the circadian regulator PER2.
PloS one (2017-05-26)
Gracie Wee Ling Eng, Edison, David M Virshup
ZUSAMMENFASSUNG
Circadian rhythms are intrinsic ~24 hour cycles that regulate diverse aspects of physiology, and in turn are regulated by interactions with the external environment. Casein kinase 1 delta (CK1δ, CSNK1D) is a key regulator of the clock, phosphorylating both stabilizing and destabilizing sites on the PER2 protein, in a mechanism known as the phosphoswitch. CK1δ can itself be regulated by phosphorylation on its regulatory domain, but the specific sites involved, and the role this plays in control of circadian rhythms as well as other CK1-dependent processes is not well understood. Using a sensitized PER2::LUC reporter assay, we identified a specific phosphorylation site, T347, on CK1δ, that regulates CK1δ activity towards PER2. A mutant CK1δ T347A was more active in promoting PER2 degradation. This CK1δ regulatory site is phosphorylated in cells in trans by dinaciclib- and staurosporine-sensitive kinases, consistent with their potential regulation by cyclin dependent and other proline-directed kinases. The regulation of CK1δ by site-specific phosphorylation via the cell cycle and other signaling pathways provides a mechanism to couple external stimuli to regulation of CK1δ-dependent pathways including the circadian clock.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Myc-Tag-Antikörper, Klon 4A6, clone 4A6, Upstate®, from mouse