- Use of pulsed-field gel electrophoresis for epidemiological study of Escherichia coli O157:H7 during a food-borne outbreak.
Use of pulsed-field gel electrophoresis for epidemiological study of Escherichia coli O157:H7 during a food-borne outbreak.
Applied and environmental microbiology (1995-07-01)
J M Johnson, S D Weagant, K C Jinneman, J L Bryant
PMID7618896
ZUSAMMENFASSUNG
Food and patient isolates from an Escherichia coli O157:H7 outbreak associated with undercooked ground beef were characterized by pulsed-field gel electrophoresis and Shiga-like toxin genotype. Pulsed-field gel electrophoresis confirmed the epidemiologically implicated source of the two-state outbreak and differentiated between outbreak and sporadic strains.