Merck
  • Matrix metalloproteinase 12 expression is associated with tumor FOXP3+ regulatory T cell infiltration and poor prognosis in hepatocellular carcinoma.
Matrix metalloproteinase 12 expression is associated with tumor FOXP3+ regulatory T cell infiltration and poor prognosis in hepatocellular carcinoma.
Oncology letters (2018-06-22)
Min-Ke He, Yong Le, Yong-Fa Zhang, Han-Yue Ouyang, Pei-En Jian, Zi-Shan Yu, Li-Juan Wang, Ming Shi
ZUSAMMENFASSUNG
Hepatocellular carcinoma (HCC) is among the most fatal types of cancer worldwide due to its high rates of recurrence and metastasis. The molecular processes involved in HCC progression require further investigation to identify biomarkers for use in diagnosis and treatment. In the present study, the significance and prognostic value of matrix metallopeptidase 12 (MMP12) expression in human HCC was investigated. MMP12 mRNA expression was investigated using reverse transcription-quantitative polymerase chain reaction analysis of 42 pairs of tumor and non-tumor liver tissues obtained from patients with HCC following surgical treatment. Immunohistochemical staining was used to detect MMP12 and forkhead box P3 (FOXP3) expression in 158 paraffin-embedded HCC tissues. The prognostic value of MMP12 expression was determined using Kaplan-Meier analysis and the Cox proportional hazards model. MMP12 mRNA levels were significantly higher in liver tumor tissues compared with matched non-tumor liver tissues. MMP12 expression and FOXP3+ regulatory T cell (Treg) infiltration was positively correlated (r=0.302; P<0.001). MMP12 protein overexpression was positively correlated with tumor size (P=0.018), high serum alpha-fetoprotein levels (P=0.005) and poor overall survival time (P=0.012) in patients with HCC. Furthermore, MMP12 protein level was an independent predictive factor for overall survival time of patients with HCC who underwent curative resection. In conclusion, these results suggest that MMP12 may increase FOXP3+ Treg infiltration into tumor tissues, and promote tumor progression and immune evasion of HCC. The overexpression of MMP12 protein is, therefore, a valuable prognostic indicator in patients with HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Normale Kaninchen-IgG-Antikörper, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.