Merck
p53CSV, a novel p53-inducible gene involved in the p53-dependent cell-survival pathway.
Cancer research (2005-03-01)
Woong-Ryeon Park, Yusuke Nakamura
ZUSAMMENFASSUNG
Although a number of p53 target genes have been identified, the mechanisms of p53-dependent activities that determine cellular survival or death are still not fully understood. Here we report isolation of a novel p53 target gene, designated p53-inducible cell-survival factor (p53CSV). p53CSV contains a p53-binding site within its second exon and the reduction of expression by small interfering RNA enhanced apoptosis, whereas overexpression protected cells from apoptosis caused by DNA damage. p53CSV is induced significantly when cells have a low level of genotoxic stresses, but not when DNA damage is severe. p53CSV can modulate apoptotic pathways through interaction with Hsp70 that probably inhibits activity of apoptosis protease activating factor-1. Our results imply that under specific conditions of stress, p53 regulates transcription of p53CSV and that p53CSV is one of the important players in the p53-mediated cell survival.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Monoclonal Anti-p53 in Maus hergestellte Antikörper, clone DO-7, purified from hybridoma cell culture