Merck
Mapping axon initial segment structure and function by multiplexed proximity biotinylation.
Nature communications (2020-01-05)
Hamdan Hamdan, Brian C Lim, Tomohiro Torii, Abhijeet Joshi, Matthias Konning, Cameron Smith, Donna J Palmer, Philip Ng, Christophe Leterrier, Juan A Oses-Prieto, Alma L Burlingame, Matthew N Rasband
ZUSAMMENFASSUNG
Axon initial segments (AISs) generate action potentials and regulate the polarized distribution of proteins, lipids, and organelles in neurons. While the mechanisms of AIS Na+ and K+ channel clustering are understood, the molecular mechanisms that stabilize the AIS and control neuronal polarity remain obscure. Here, we use proximity biotinylation and mass spectrometry to identify the AIS proteome. We target the biotin-ligase BirA* to the AIS by generating fusion proteins of BirA* with NF186, Ndel1, and Trim46; these chimeras map the molecular organization of AIS intracellular membrane, cytosolic, and microtubule compartments. Our experiments reveal a diverse set of biotinylated proteins not previously reported at the AIS. We show many are located at the AIS, interact with known AIS proteins, and their loss disrupts AIS structure and function. Our results provide conceptual insights and a resource for AIS molecular organization, the mechanisms of AIS stability, and polarized trafficking in neurons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-GAPDH in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Tubulin-Antikörper, Beta-III-Isoform, C-Terminus, Klon TU-20 (TUJ1 ähnlich), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®