Merck
  • In Vitro Evidence for Effects of Magnesium Supplementation on Quinolone-treated Horse and Dog Chondrocytes
<I>In Vitro</I> Evidence for Effects of Magnesium Supplementation on Quinolone-treated Horse and Dog Chondrocytes
Veterinary Pathology (2001)
M. Egerbacher, B. Wolfesberger, et al.