Merck
  • Single cell census of human kidney organoids shows reproducibility and diminished off-target cells after transplantation.
Single cell census of human kidney organoids shows reproducibility and diminished off-target cells after transplantation.
Nature communications (2019-12-01)
Ayshwarya Subramanian, Eriene-Heidi Sidhom, Maheswarareddy Emani, Katherine Vernon, Nareh Sahakian, Yiming Zhou, Maria Kost-Alimova, Michal Slyper, Julia Waldman, Danielle Dionne, Lan T Nguyen, Astrid Weins, Jamie L Marshall, Orit Rosenblatt-Rosen, Aviv Regev, Anna Greka
ZUSAMMENFASSUNG
Human iPSC-derived kidney organoids have the potential to revolutionize discovery, but assessing their consistency and reproducibility across iPSC lines, and reducing the generation of off-target cells remain an open challenge. Here, we profile four human iPSC lines for a total of 450,118 single cells to show how organoid composition and development are comparable to human fetal and adult kidneys. Although cell classes are largely reproducible across time points, protocols, and replicates, we detect variability in cell proportions between different iPSC lines, largely due to off-target cells. To address this, we analyze organoids transplanted under the mouse kidney capsule and find diminished off-target cells. Our work shows how single cell RNA-seq (scRNA-seq) can score organoids for reproducibility, faithfulness and quality, that kidney organoids derived from different iPSC lines are comparable surrogates for human kidney, and that transplantation enhances their formation by diminishing off-target cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin in Kaninchen hergestellte Antikörper, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution