Merck
  • An in situ hybridization technique to detect low-abundance slug mRNA in adherent cultured cells.
An in situ hybridization technique to detect low-abundance slug mRNA in adherent cultured cells.
Methods in molecular biology (Clifton, N.J.) (2006-06-20)
Changsun Choi, Laurie G Hudson, Pierre Savagner, Donna F Kusewitt
ZUSAMMENFASSUNG
In this chapter, we describe a simple and relatively rapid technique for detecting low-abundance slug mRNA in cultured cells. The procedure uses nonradioactive digoxigenin-labeled RNA probes that are more sensitive than deoxyribonucleic probes and simpler to detect than radioactively labeled probes. Cells are grown in glass chamber slides, fixed in an acidic fixative, dehydrated through ethanol and xylene, permeabilized in pepsin, and post-fixed. Slides are then incubated overnight at 37 degrees C in a buffer containing 50% formamide and 5-10 ng/gL probe. Excess probe is removed by washing at high temperature in low-salt buffer and by treatment with RNase. Probe is detected immunohistochemically with an anti-digoxigenin Fab fragment, using a tyramide amplification kit to enhance signal and Fast Red for visualization. The technique has the advantages of probe stability and sensitivity, hybridization at low temperature, rapidity and sensitivity of probe detection, and the production of permanent specimens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ribonukleinsäure, Transfer aus Backhefe (S. cerevisiae), buffered aqueous solution