Merck
  • Immunofluorescence Labeling of a Mutant of Tissue Non-Specific Alkaline Phosphatase Lacking the Glysosylphosphatidylinositol Anchor.
Immunofluorescence Labeling of a Mutant of Tissue Non-Specific Alkaline Phosphatase Lacking the Glysosylphosphatidylinositol Anchor.
Journal of Nippon Medical School = Nippon Ika Daigaku zasshi (2016-09-30)
Tomohiro Matsumura, Shiho Saito, Hideo Orimo
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-ALPL antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution