Merck
  • Schizophrenia-related neural and behavioral phenotypes in transgenic mice expressing truncated Disc1.
Schizophrenia-related neural and behavioral phenotypes in transgenic mice expressing truncated Disc1.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2008-10-24)
Sanbing Shen, Bing Lang, Chizu Nakamoto, Feng Zhang, Jin Pu, Soh-Leh Kuan, Christina Chatzi, Shuisheng He, Iain Mackie, Nicholas J Brandon, Karen L Marquis, Mark Day, Orest Hurko, Colin D McCaig, Gernot Riedel, David St Clair
ZUSAMMENFASSUNG
Disrupted-in-Schizophrenia-1 (DISC1), identified by positional cloning of a balanced translocation (1;11) with the breakpoint in intron 8 of a large Scottish pedigree, is associated with a range of neuropsychiatric disorders including schizophrenia. To model this mutation in mice, we have generated Disc1(tr) transgenic mice expressing 2 copies of truncated Disc1 encoding the first 8 exons using a bacterial artificial chromosome (BAC). With this partial simulation of the human situation, we have discovered a range of phenotypes including a series of novel features not previously reported. Disc1(tr) transgenic mice display enlarged lateral ventricles, reduced cerebral cortex, partial agenesis of the corpus callosum, and thinning of layers II/III with reduced neural proliferation at midneurogenesis. Parvalbumin GABAergic neurons are reduced in the hippocampus and medial prefrontal cortex, and displaced in the dorsolateral frontal cortex. In culture, transgenic neurons grow fewer and shorter neurites. Behaviorally, transgenic mice exhibit increased immobility and reduced vocalization in depression-related tests, and impairment in conditioning of latent inhibition. These abnormalities in Disc1(tr) transgenic mice are consistent with findings in severe schizophrenia.