Merck
  • Molecular and cytogenetic characterization of repetitive DNA in the Antarctic polyplacophoran Nuttallochiton mirandus.
Molecular and cytogenetic characterization of repetitive DNA in the Antarctic polyplacophoran Nuttallochiton mirandus.
Chromosome research : an international journal on the molecular, supramolecular and evolutionary aspects of chromosome biology (2008-08-06)
Maria Assunta Biscotti, Marco Barucca, Teresa Capriglione, Gaetano Odierna, Ettore Olmo, Adriana Canapa
ZUSAMMENFASSUNG
Two highly repeated DNAs, designated NmE1/NmE2 and NmE5, were identified by EcoRV digestion in the chiton Nuttallochiton mirandus (Mollusca: Polyplacophora). The comparison of the sequences obtained showed high similarity in 5' and 3' regions and the NmE5 sequence displayed an inserted sequence that might arise from a transposable element. Southern blotting analyses suggested a tandem organization of both satellite DNA families identified. Moreover, dot blot analyses, performed on several molluscan species, revealed a different degree of conservation of the repeated DNAs. Fluorescence in-situ hybridizations (FISH) on metaphase chromosomes showed that both satellite DNAs are located at centromeric regions.