Merck
  • ITS-RFLP- and RAPD-based genetic variability of Trypanosoma cruzi I, human and vector strains in Santander, Colombia.
ITS-RFLP- and RAPD-based genetic variability of Trypanosoma cruzi I, human and vector strains in Santander, Colombia.
Parasitology research (2009-04-07)
Katherine Paola Luna-Marín, Claudia Lorena Jaramillo-Londoño, Jorge Hernández-Torres, Reynaldo Gutiérrez-Marín, Gustavo Adolfo Vallejo, Víctor Manuel Angulo-Silva
ZUSAMMENFASSUNG
Chagas disease is a severe public health problem in Latin-American countries. In Colombia, the predominance of Trypanosoma cruzi I has been described in the literature, with a broad heterogeneity between strains. However, most of the studies carried out centered on isoenzyme analysis, with a smaller number that focus on other molecular methods. In this report, we discuss the results of a molecular analysis of T. cruzi I strains, isolated from the domestic cycle, from the department of Santander, one of the territorial divisions where the prevalence of infection is highest. Internal transcribed spacer-restriction fragment length polymorphism and random amplification of polymorphic DNA were used to characterize 16 strains from human and vector (Triatominae) hosts. The data reveal a clustering based on the biological origin. Human and vector strains grouped separately; however, three vector strains clustered together with human strains. These results indicate that genetic differences exist in the strains that infect both hosts. We conclude that T. cruzi I populations in the domestic cycle of transmission of Chagas disease in Santander are heterogeneous and are composed of different clones. The epidemiological and biological implications are discussed.