Merck
  • Myelinosomes act as natural secretory organelles in Sertoli cells to prevent accumulation of aggregate-prone mutant Huntingtin and CFTR.
Myelinosomes act as natural secretory organelles in Sertoli cells to prevent accumulation of aggregate-prone mutant Huntingtin and CFTR.
Human molecular genetics (2016-08-06)
Marina G Yefimova, Emile Béré, Anne Cantereau-Becq, Thomas Harnois, Annie-Claire Meunier, Nadia Messaddeq, Frédéric Becq, Yvon Trottier, Nicolas Bourmeyster
ZUSAMMENFASSUNG
Inappropriate deposition of insoluble aggregates of proteins with abnormal structures is a hallmark of affected organs in protein aggregation disease. Very rare, affected organs avoid aggregation naturally. This concerns atrophic testis in Huntington disease (HD). We aimed to understand how HD testis avoids aggregation. Using HD model R6/1 mice, we demonstrate that affected testis contain rare organelles myelinosomes. Myelinosomes secreted from testis somatic TM4 Sertoli cells provide the release of aggregate-prone mutant, but not normal Huntingtin (Htt) exon1. Myelinosomes also support the release of other aggregate-prone mutant protein responsible for cystic fibrosis (CF), F508delCFTR. The traffic and discharge of myelinosomes is facilitated by multivesicular bodies (MVB)s. Inhibition of MVB excretion induced reversible retention of both misfolded proteins inside TM4 Sertoli cells. We propose that myelinosome-mediated elimination of mutant proteins is an unusual secretory process allowing Sertoli cells getting rid of misfolded proteins to avoid aggregation and to maintain cell proteostasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
(−)-Blebbistatin, solid, synthetic
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)