Merck
  • Improvement of visual performance with intravitreal administration of 9-cis-retinal in Rpe65-mutant dogs.
Improvement of visual performance with intravitreal administration of 9-cis-retinal in Rpe65-mutant dogs.
Archives of ophthalmology (Chicago, Ill. : 1960) (2010-09-15)
Patricia M Gearhart, Chris Gearhart, Debra A Thompson, Simon M Petersen-Jones
ZUSAMMENFASSUNG
To determine the efficacy of intravitreal administration of 9-cis-retinal in restoring visual function in Rpe65-mutant dogs. Intravitreal injection of 9-cis-retinal was administered in 1 eye of 7 Rpe65-/- dogs at a range of ages. Electroretinogram analysis and testing of visual performance was used to evaluate outcomes after a single injection and in 2 dogs after a second injection in the same eye. In 5 of 7 injected dogs, 9-cis-retinal injection resulted in increased rod electroretinogram responses and improved functional vision. Three injected dogs exhibited increased 33-Hz flicker amplitudes characteristic of cone-mediated responses. Electroretinogram improvement was no longer evident by week 10 postinjection in 1 dog monitored over time. A second injection of 9-cis-retinal was performed in the same eye of 2 of the 7 dogs and also resulted in rescue of visual function. Our findings establish that 9-cis-retinoid therapy can restore visual function in a canine model of human disease resulting from RPE65 mutations. These positive proof-of-principle results provide support for the development of intravitreal devices for sustained delivery of 9-cis-retinal as a therapy for conditions resulting from failure of the visual cycle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9-cis-Retinal, vitamin A analog