- A human induced pluripotent stem cell line (TRNDi007-B) from an infantile onset Pompe patient carrying p.R854X mutation in the GAA gene.
A human induced pluripotent stem cell line (TRNDi007-B) from an infantile onset Pompe patient carrying p.R854X mutation in the GAA gene.
Stem cell research (2019-04-27)
Yu-Shan Cheng, Rong Li, Amanda Baskfield, Jeanette Beers, Jizhong Zou, Chengyu Liu, Wei Zheng
PMID31026687
ZUSAMMENFASSUNG
Pompe disease is an autosomal inherent genetic disease caused by mutations in the GAA gene that encodes acid alpha-glucosidase. The disease affects patients in heart, skeletal muscles, liver, and central nervous system. A human induced pluripotent stem cell (iPSC) line was generated from the skin dermal fibroblasts of a Pompe patient with homozygosity for a c.2560C > T (p.R854X) mutation in exon 18 of the GAA gene. This human iPSC line provides a useful resource for disease modeling and drug discovery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit