Merck
  • Hydrophobic interaction chromatography on octyl sepharose--an approach for one-step platform purification of cyclodextrin glucanotransferases.
Hydrophobic interaction chromatography on octyl sepharose--an approach for one-step platform purification of cyclodextrin glucanotransferases.
Preparative biochemistry & biotechnology (2011-10-05)
Premalatha Shetty, Smitha Bhat, J L Iyer, Srikant Shenoy, J S Pai, K Satyamoorthy
ZUSAMMENFASSUNG
Cyclodextrin glucanotransferase (CGTase) from Bacillus circulans ATCC 21783 was concentrated by ultrafiltration and subsequently purified by hydrophobic interaction chromatography on Octyl Sepharose 4 fast flow. The matrix was able to bind selectively to the enzyme at a very low ammonium sulfate concentration of 0.67 M and enzyme desorption was performed by decreasing gradient of the salt. The overall recovery was 80% with 689-fold purity. CGTases derived from four soil isolates and Toruzyme, the commercial preparation of CGTase, also bound to Octyl Sepharose under similar conditions at 0.67 M and eluted at 0.55-0.5 M of ammonium sulfate. Octyl Sepharose chromatography can thus be used as a platform approach for purification of CGTases from various bacterial sources. Long stretches of sequence predominated by hydrophobic amino acids are reportedly present in the starch binding domains of CGTases. Starch binding experiments indicated the binding of the enzymes to the octyl matrix through these domains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Octyl-Sepharose CL-4B