Merck
  • Predicting puberty in partial androgen insensitivity syndrome: Use of clinical and functional androgen receptor indices.
Predicting puberty in partial androgen insensitivity syndrome: Use of clinical and functional androgen receptor indices.
EBioMedicine (2018-10-15)
Ngee Lek, Rieko Tadokoro-Cuccaro, Jonathan B Whitchurch, Bismoy Mazumder, Harriet Miles, Philippa Prentice, Trevor Bunch, Karolina Zielińska, Veronika Metzler, Nigel P Mongan, David M Heery, Ieuan A Hughes
ZUSAMMENFASSUNG
PAIS exhibits a complex spectrum of phenotypes and pubertal outcomes. The paucity of reliable prognostic indicators can confound management decisions including sex-of-rearing. We assessed whether external masculinisation score (EMS) at birth or functional assays correlates with pubertal outcome in PAIS patients and whether the EMS is helpful in sex assignment. We collected pubertal outcome data for 27 male-assigned PAIS patients, all with confirmed androgen receptor (AR) mutations, including two previously uncharacterized variants (I899F; Y916C). Patients were grouped as follows; EMS at birth <5 and ≥ 5 (EMS in normal males is 12; median EMS in PAIS is 4·7) and pubertal outcomes compared. Only 6/9 patients (67%) with EMS <5 underwent spontaneous onset of puberty, versus all 18 patients with EMS ≥5 (p = .03). Only 1/6 patients (17%) with EMS <5 developed adult genitalia reaching Tanner stage 4 or 5, versus 11/13 (85%) with EMS ≥5 (p = 0·01). There was no significant difference between the two groups of patients in being prescribed androgen replacement, who reached adult testicular volume ≥ 15 ml, pubic hair Tanner stage 4 or 5, above average adult height, had gynaecomastia, and mastectomy. No correlation was observed between EMS and in vitro AR function. In PAIS with AR mutation, birth EMS is a simple predictor of spontaneous pubertal onset and satisfactory adult genitalia. This provides useful information when discussing the likely options for management at puberty. FUND: European Commission Framework 7 Programme, NIHR Cambridge Biomedical Research Centre, BBSRC DTP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-LexA-Antikörper, DNA-Bindungsregion, Upstate®, from rabbit