Merck
  • Insertion of channel of phi29 DNA packaging motor into polymer membrane for high-throughput sensing.
Insertion of channel of phi29 DNA packaging motor into polymer membrane for high-throughput sensing.
Nanomedicine : nanotechnology, biology, and medicine (2020-02-09)
Zhouxiang Ji, Michael Jordan, Lakmal Jayasinghe, Peixuan Guo
ZUSAMMENFASSUNG
The connector channel of bacteriophage phi29 DNA packaging motor has been inserted into the lipid bilayer membrane and has shown potential for the sensing of DNA, RNA, chemicals, peptides, and antibodies. Properties such as high solubility and large channel size have made phi29 channel an advantageous system for those applications; however, previously studied lipid membranes have short lifetimes, and they are frangible and unstable under voltages higher than 200 mV. Thus, the application of this lipid membrane platform for clinical applications is challenging. Here we report the insertion of the connector into the stable polymer membrane in MinION flow cell that contains 2048 wells for high-throughput sensing by the liposome-polymer fusion process. The successful insertion of phi29 connector was confirmed by a unique gating phenomenon. Peptide translocation through the inserted phi29 connector was also observed, revealing the potential of applying phi29 connector for high-throughput peptide sensing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand
Avanti
4ME 16:0 PC, Avanti Research - A Croda Brand