Merck
  • Mechanical behavior and microstructure of cement composites incorporating surface-treated multi-walled carbon nanotubes
Mechanical behavior and microstructure of cement composites incorporating surface-treated multi-walled carbon nanotubes
Carbon (2005)
Li GY, et al.