Merck
  • A novel mode for integrin-mediated signaling: tethering is required for phosphorylation of FAK Y397.
A novel mode for integrin-mediated signaling: tethering is required for phosphorylation of FAK Y397.
Molecular biology of the cell (2003-09-10)
Qi Shi, David Boettiger
ZUSAMMENFASSUNG
The common model for integrin mediated signaling is based on integrin clustering and the potential for that clustering to recruit signaling molecules including FAK and src. The clustering model for transmembrane signaling originated with the analysis of the EGF receptor signaling and remains the predominant model. The roles for substrate-bound ligand and ligand occupancy in integrin-mediated signaling are less clear. A kinetic model was established using HT1080 cells in which there was a linear relationship between the strength of adhesion, the proportion of alpha5beta1 integrin that could be chemically cross-linked, and the number of receptor-ligand bonds. This graded signal produced a similarly graded response measured by the level of specific phosphorylation of FAK Y397. FAK Y397 phosphorylation could also be induced by antibody bound to the substrate. In contrast, clustering of alpha5beta1 on suspended cells with either antibody to beta1 or by clustering of soluble ligand bound to alpha5beta1 induced the phosphorylation of FAK Y861 but not Y397. There were no differences in signaling when activating antibodies were compared with blocking antibodies, presence or absence of ligand. Only tethering of alpha5beta1 to the substrate was required for induction of FAK Y397 phosphorylation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse