Merck
  • Monitoring mRNA Translation in Neuronal Processes Using Fluorescent Non-Canonical Amino Acid Tagging.
Monitoring mRNA Translation in Neuronal Processes Using Fluorescent Non-Canonical Amino Acid Tagging.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2016-03-31)
Aron Kos, Kai A Wanke, Anthony Gioio, Gerard J Martens, Barry B Kaplan, Armaz Aschrafi
ZUSAMMENFASSUNG
A steady accumulation of experimental data argues that protein synthesis in neurons is not merely restricted to the somatic compartment, but also occurs in several discrete cellular micro-domains. Local protein synthesis is critical for the establishment of synaptic plasticity in mature dendrites and in directing the growth cones of immature axons, and has been associated with cognitive impairment in mice and humans. Although in recent years a number of important mechanisms governing this process have been described, it remains technically challenging to precisely monitor local protein synthesis in individual neuronal cell parts independent from the soma. This report presents the utility of employing microfluidic chambers for the isolation and treatment of single neuronal cellular compartments. Furthermore, it is demonstrated that a protein synthesis assay, based on fluorescent non-canonical amino acid tagging (FUNCAT), can be combined with this cell culture system to label nascent proteins within a discrete structural and functional domain of the neuron. Together, these techniques could be employed for the detection of protein synthesis within developing and mature neurites, offering an effective approach to elucidate novel mechanisms controlling synaptic maintenance and plasticity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Tau-1-Antikörper, Klon PC1C6, clone PC1C6, Chemicon®, from mouse