Merck
miRNA-mediated TUSC3 deficiency enhances UPR and ERAD to promote metastatic potential of NSCLC.
Nature communications (2018-12-07)
Young-Jun Jeon, Taewan Kim, Dongju Park, Gerard J Nuovo, Siyeon Rhee, Pooja Joshi, Bum-Kyu Lee, Johan Jeong, Sung-Suk Suh, Jeff E Grotzke, Sung-Hak Kim, Jieun Song, Hosung Sim, Yonghwan Kim, Yong Peng, Youngtae Jeong, Michela Garofalo, Nicola Zanesi, Jonghwan Kim, Guang Liang, Ichiro Nakano, Peter Cresswell, Patrick Nana-Sinkam, Ri Cui, Carlo M Croce
ZUSAMMENFASSUNG
Non-small cell lung carcinoma (NSCLC) is leading cause of cancer-related deaths in the world. The Tumor Suppressor Candidate 3 (TUSC3) at chromosome 8p22 known to be frequently deleted in cancer is often found to be deleted in advanced stage of solid tumors. However, the role of TUSC3 still remains controversial in lung cancer and context-dependent in several cancers. Here we propose that miR-224/-520c-dependent TUSC3 deficiency enhances the metastatic potential of NSCLC through the alteration of three unfolded protein response pathways and HRD1-dependent ERAD. ATF6α-dependent UPR is enhanced whereas the affinity of HRD1 to its substrates, PERK, IRE1α and p53 is weakened. Consequently, the alteration of UPRs and the suppressed p53-NM23H1/2 pathway by TUSC3 deficiency is ultimately responsible for enhancing metastatic potential of lung cancer. These findings provide mechanistic insight of unrecognized roles of TUSC3 in cancer progression and the oncogenic role of HRD1-dependent ERAD in cancer metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-TUSC3 antibody produced in rabbit, affinity isolated antibody