Merck
  • Tight junction protein claudin-1 enhances the invasive activity of oral squamous cell carcinoma cells by promoting cleavage of laminin-5 gamma2 chain via matrix metalloproteinase (MMP)-2 and membrane-type MMP-1.
Tight junction protein claudin-1 enhances the invasive activity of oral squamous cell carcinoma cells by promoting cleavage of laminin-5 gamma2 chain via matrix metalloproteinase (MMP)-2 and membrane-type MMP-1.
Cancer research (2006-05-19)
Naohisa Oku, Eri Sasabe, Eisaku Ueta, Tetsuya Yamamoto, Tokio Osaki
ZUSAMMENFASSUNG
Although adherent junctions have been extensively studied, the role of tight junctions in cancer cell invasion is not sufficiently explored. We investigated whether claudin-1, a component of tight junctions, regulated invasion activity in oral squamous cell carcinoma (OSC) cells. The expression of claudin-1, activity of matrix metalloproteinase (MMP)-2, and cleavage of laminin-5 gamma2 chains were assessed by Western blot analysis, immunohistochemistry, and zymography in OSC cell lines (OSC-4 and NOS-2, highly invasive; OSC-7, weakly invasive) and their xenografts in severe combined immunodeficient (SCID) mice. The influence of claudin-1 small interfering RNA (siRNA) on the invasion activity of the cell lines was also investigated. Compared with OSC-7, both OSC-4 and NOS-2 more strongly expressed claudin-1 and possessed high activities of MMP-2 and MMP-9. Tumors formed in the tongues of SCID mice xenografted with OSC-4, NOS-2, and OSC-7 immunohistochemically revealed strong, moderate, and weak expression of laminin-5 gamma2 chains, respectively, and laminin-5 gamma2 chains were secreted in the conditioned medium of the cancer cells in parallel with the in vivo results. Claudin-1 siRNA largely suppressed the invasion of OSC-4 and decreased the activation of MMP-2, the expression of membrane-type MMP-1 (MT1-MMP), and the cleavage of laminin-5 gamma2. In addition, not only antibodies against MT1-MMP and epidermal growth factor receptor (EGFR) but also MMP-2 and EGFR inhibitors strongly suppressed the invasion activity of OSC-4. These results suggest that claudin-1 up-regulates cancer cell invasion activity through activation of MT1-MMP and MMP-2, which results in enhanced cleavage of laminin-5 gamma2 chains.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Laminin-5-(γ2-Kette-)Antikörper, Klon D4B5, clone D4B5, Chemicon®, from mouse
Sigma-Aldrich
Anti-EGFR-Antikörper, neutralisierend, Klon LA1, clone LA1, Upstate®, from mouse