Merck
  • Thioredoxin-2 inhibits mitochondrial reactive oxygen species generation and apoptosis stress kinase-1 activity to maintain cardiac function.
Thioredoxin-2 inhibits mitochondrial reactive oxygen species generation and apoptosis stress kinase-1 activity to maintain cardiac function.
Circulation (2015-01-30)
Qunhua Huang, Huanjiao Jenny Zhou, Haifeng Zhang, Yan Huang, Ford Hinojosa-Kirschenbaum, Peidong Fan, Lina Yao, Luiz Belardinelli, George Tellides, Frank J Giordano, Grant R Budas, Wang Min
ZUSAMMENFASSUNG
Thioredoxin 2 (Trx2) is a key mitochondrial protein that regulates cellular redox and survival by suppressing mitochondrial reactive oxygen species generation and by inhibiting apoptosis stress kinase-1 (ASK1)-dependent apoptotic signaling. To date, the role of the mitochondrial Trx2 system in heart failure pathogenesis has not been investigated. Western blot and histological analysis revealed that Trx2 protein expression levels were reduced in hearts from patients with dilated cardiomyopathy, with a concomitant increase in ASK1 phosphorylation/activity. Cardiac-specific Trx2 knockout mice develop spontaneous dilated cardiomyopathy at 1 month of age with increased heart size, reduced ventricular wall thickness, and a progressive decline in left ventricular contractile function, resulting in mortality due to heart failure by ≈4 months of age. The progressive decline in cardiac function observed in cardiac-specific Trx2 knockout mice was accompanied by the disruption of mitochondrial ultrastructure, mitochondrial membrane depolarization, increased mitochondrial reactive oxygen species generation, and reduced ATP production, correlating with increased ASK1 signaling and increased cardiomyocyte apoptosis. Chronic administration of a highly selective ASK1 inhibitor improved cardiac phenotype and reduced maladaptive left ventricular remodeling with significant reductions in oxidative stress, apoptosis, fibrosis, and cardiac failure. Cellular data from Trx2-deficient cardiomyocytes demonstrated that ASK1 inhibition reduced apoptosis and reduced mitochondrial reactive oxygen species generation. Our data support an essential role for mitochondrial Trx2 in preserving cardiac function by suppressing mitochondrial reactive oxygen species production and ASK1-dependent apoptosis. Inhibition of ASK1 represents a promising therapeutic strategy for the treatment of dilated cardiomyopathy and heart failure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Cre-Rekombinase-Antikörper, Klon 2D8, ascites fluid, clone 2D8, Chemicon®
Sigma-Aldrich
Anti-Mitochondrien-Antikörper, Oberfläche intakter Mitochondrien, Klon 113-1, clone 113-1, Chemicon®, from mouse