Merck
  • A new role for the Kruppel-like transcription factor KLF6 as an inhibitor of c-Jun proto-oncoprotein function.
A new role for the Kruppel-like transcription factor KLF6 as an inhibitor of c-Jun proto-oncoprotein function.
Oncogene (2004-09-21)
Daniela A Slavin, Nicolás P Koritschoner, Claudio C Prieto, Fernando J López-Díaz, Bruno Chatton, José Luis Bocco
ZUSAMMENFASSUNG
Kruppel-like transcription factors (KLFs) represent one of the most diverse set of regulators in vertebrate organisms. KLF family members are involved in cell proliferation and differentiation control in normal as well as in pathological situations. Here, we demonstrate that KLF6 behaves as a functional antagonist of the c-Jun proto-oncoprotein. Thus, KLF6 overexpression downregulated c-Jun-dependent transcription and a physical interaction between c-Jun and KLF6 was detected. Moreover, cell proliferation induced by c-Jun was significantly decreased by KLF6. The inhibition of c-Jun functions correlates directly with c-Jun protein degradation induced by KLF6. We also show that all KLF6 effects on c-Jun were largely dependent on phorbol ester (TPA/ionomycin) extracellular stimulation, which enhanced KLF6 nuclear translocation and transcriptional activity and modified its phosphorylation status. Our data are consistent with a novel mechanism of KLF6's role as an inhibitor of cell proliferation by counteracting the function of the c-Jun proto-oncoprotein involving enhanced c-Jun degradation by the proteasome-dependent pathway, and further reinforces KLF6 as a potential tumor suppressor gene product.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-Phosphoserin, clone PSR-45, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Phosphothreonine antibody produced in mouse, clone PTR-8, ascites fluid