Merck
Trafficking of cholesterol to the ER is required for NLRP3 inflammasome activation.
The Journal of cell biology (2018-07-29)
Marianne de la Roche, Claire Hamilton, Rebecca Mortensen, A Arockia Jeyaprakash, Sanjay Ghosh, Paras K Anand
ZUSAMMENFASSUNG
Cellular lipids determine membrane integrity and fluidity and are being increasingly recognized to influence immune responses. Cellular cholesterol requirements are fulfilled through biosynthesis and uptake programs. In an intricate pathway involving the lysosomal cholesterol transporter NPC1, the sterol gets unequally distributed across intracellular compartments. By using pharmacological and genetic approaches targeting NPC1, we reveal that blockade of cholesterol trafficking through the late endosome-lysosome pathway blunts NLRP3 inflammasome activation. Altered cholesterol localization at the plasma membrane (PM) in Npc1-/- cells abrogated AKT-mTOR signaling by TLR4. However, the inability to activate the NLRP3 inflammasome was traced to perturbed cholesterol trafficking to the ER but not the PM. Accordingly, acute cholesterol depletion in the ER membranes by statins abrogated casp-1 activation and IL-1β secretion and ablated NLRP3 inflammasome assembly. By contrast, assembly and activation of the AIM2 inflammasome progressed unrestricted. Together, this study reveals ER sterol levels as a metabolic rheostat for the activation of the NLRP3 inflammasome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenElute Miniprep-Kit für genomische Säugetier-DNA, sufficient for 70 purifications
Sigma-Aldrich
U18666A, CAS-Nr. 3039-71-2, Calbiochem, A cell-permeable, amphiphilic amino-steroid that alters intracellular membrane protein trafficking by impairing intracellular biosynthesis and transport of LDL-derived cholesterol.
Sigma-Aldrich
Anti-ASC-Antikörper, Klon 2EI-7, clone 2EI-7, from mouse