Merck
  • EZH2 Methyltransferase Activity Controls Pten Expression and mTOR Signaling during Fear Memory Reconsolidation.
EZH2 Methyltransferase Activity Controls Pten Expression and mTOR Signaling during Fear Memory Reconsolidation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2018-07-22)
Timothy J Jarome, Gabriella A Perez, Rebecca M Hauser, Katrina M Hatch, Farah D Lubin
ZUSAMMENFASSUNG
Memory retrieval induces a transient period of increased transcriptional and translational regulation in neurons called reconsolidation, which is regulated by the protein kinase B (AKT)-mammalian target of rapamycin (mTOR) pathway. However, it is currently unknown how activation of the AKT-mTOR pathway is regulated during the reconsolidation process. Here, we found that in male rats retrieval of a contextual fear memory transiently increased Enhancer of Zeste Homolog 2 (EZH2) levels along with increased histone H3 lysine 27 trimethylation (H3K27me3) levels, which correlated with decreased levels of phosphatase and tensin homolog (PTEN), a potent inhibitor of AKT-mTOR-dependent signaling in the hippocampus. Further experiments found increased H3K27me3 levels and DNA methylation across the Pten promoter and coding regions, indicating transcriptional silencing of the Pten gene. Pten H3K27me3 levels did not change following training or after the retrieval of a remote (old) fear memory, suggesting that this mechanism of Pten repression was specific to the reconsolidation of a new memory. In vivo siRNA-mediated knockdown of Ezh2 in the hippocampus abolished retrieval-induced increases in H3K27me3 and prevented decreases in PTEN levels. Ezh2 knockdown attenuated increases in the phosphorylation of AKT and mTOR following retrieval, which could be restored by simultaneously reducing Pten, suggesting that H3K27me3 regulates AKT-mTOR phosphorylation via repression of Pten Consistent with these results, knockdown of Ezh2 in area CA1 before retrieval impaired memory on later tests. Collectively, these results suggest that EZH2-mediated H3K27me3 plays a critical role in the repression of Pten transcription necessary for AKT-mTOR activation and memory reconsolidation following retrieval.SIGNIFICANCE STATEMENT Understanding how critical translation pathways, like mTOR-mediated protein synthesis, are regulated during the memory storage process is necessary for improving memory impairments. This study tests whether mTOR activation is coupled to epigenetic mechanisms in the hippocampus following the retrieval of a contextual fear memory. Specifically, this study evaluates the role of epigenetic modifications in the form of histone methylation in downstream mTOR translational control during learning-dependent synaptic plasticity in neurons. Considering the broad implications of transcriptional and translational mechanisms in synaptic plasticity, psychiatric, and neurological and neurodegenerative disorders, these data are of interest to the neuroscience community due to the robust and specific regulation of mTOR signaling we found to be dependent on repressive histone methylation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antikörper, Upstate®, from rabbit