Merck
Expression of a novel versican variant in dorsal root ganglia from spared nerve injury rats.
Molecular pain (2019-08-21)
Oliver Bogen, Olaf Bender, Pedro Alvarez, Marie Kern, Stefan Tomiuk, Ferdinand Hucho, Jon D Levine
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Versican-Antikörper, a.a. 535-598 Maus-Versican, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Versican-Antikörper, Klon 8D8.1, clone 8D8.1, from mouse