Merck
  • Caspase cleavage of the amyloid precursor protein is prevented after overexpression of bcl-2 in a triple transgenic mouse model of Alzheimer's disease.
Caspase cleavage of the amyloid precursor protein is prevented after overexpression of bcl-2 in a triple transgenic mouse model of Alzheimer's disease.
International journal of physiology, pathophysiology and pharmacology (2009-01-12)
Debra K Kumasaka, Veronica Galvan, Elizabeth Head, Troy T Rohn
ZUSAMMENFASSUNG
A recent study demonstrated the lack of beta-amyloid (Abeta) plaque formation and accumulation of the amyloid precursor protein (APP) in a triple transgenic mouse model of Alzheimer's disease (3xTg-AD) following overexpression of the anti-apoptotic protein, Bcl-2 (Rohn et al., J. Neurosci. 28: 3051-9, 2008). The supposition from that study was the accumulation of APP resulted from a decrease in caspase-mediated processing of APP. To determine a direct role for the caspase-cleavage of APP in 3xTg-AD mice, we designed a site-directed caspase-cleavage antibody to APP and demonstrated it is a specific marker for caspase-cleaved APP. Application of this antibody revealed neuronal staining in the hippocampus and subiculum of 3xTg-AD mice. These results were confirmed utilizing a similar site-directed antibody to caspase-cleaved APP (APPneo). The caspase cleavage of APP as well as the formation of extracellular Abeta plaques was prevented in 3xTg-AD animals overexpressing Bcl-2. These results provide further support that caspases play a proximal role in promoting the pathology associated with AD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Tau Antibody, Caspase Cleaved (truncated at Asp421), clone tau-C3, Chemicon®, from mouse