Merck
Kindlin-2 controls bidirectional signaling of integrins.
Genes & development (2008-05-17)
Eloi Montanez, Siegfried Ussar, Martina Schifferer, Michael Bösl, Roy Zent, Markus Moser, Reinhard Fässler
ZUSAMMENFASSUNG
Control of integrin activation is required for cell adhesion and ligand-induced signaling. Here we report that loss of the focal adhesion protein Kindlin-2 in mice results in peri-implantation lethality caused by severe detachment of the endoderm and epiblast from the basement membrane. We found that Kindlin-2-deficient cells were unable to activate their integrins and that Kindlin-2 is required for talin-induced integrin activation. Furthermore, we demonstrate that Kindlin-2 is required for integrin outside-in signaling to enable firm adhesion and spreading. Our findings provide evidence that Kindlin-2 is a novel and essential element of bidirectional integrin signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Integrin-β-1-Antikörper, Klon MB1.2, clone MB1.2, Chemicon®, from rat