Merck
  • Physical and functional interactions between a glioma cation channel and integrin-β1 require α-actinin.
Physical and functional interactions between a glioma cation channel and integrin-β1 require α-actinin.
American journal of physiology. Cell physiology (2015-06-26)
Arun K Rooj, Zhiyong Liu, Carmel M McNicholas, Catherine M Fuller
ZUSAMMENFASSUNG
Major plasma membrane components of the tumor cell, ion channels, and integrins play crucial roles in metastasis. Glioma cells express an amiloride-sensitive nonselective cation channel composed of acid-sensing ion channel (ASIC)-1 and epithelial Na(+) channel (ENaC) α- and γ-subunits. Inhibition of this channel is associated with reduced cell migration and proliferation. Using the ASIC-1 subunit as a reporter for the channel complex, we found a physical and functional interaction between this channel and integrin-β1. Short hairpin RNA knockdown of integrin-β1 attenuated the amiloride-sensitive current, which was due to loss of surface expression of ASIC-1. In contrast, upregulation of membrane expression of integrin-β1 increased the surface expression of ASIC-1. The link between the amiloride-sensitive channel and integrin-β1 was mediated by α-actinin. Downregulation of α-actinin-1 or -4 attenuated the amiloride-sensitive current. Mutation of the putative binding site for α-actinin on the COOH terminus of ASIC-1 reduced the membrane localization of ASIC-1 and also resulted in attenuation of the amiloride-sensitive current. Our data suggest a novel interaction between the amiloride-sensitive glioma cation channel and integrin-β1, mediated by α-actinin. This interaction may form a mechanism by which channel activity can regulate glioma cell proliferation and migration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone HB1.1, clone HB1.1, Chemicon®, from mouse