Merck
Mechanical stress alters protein O-GlcNAc in human periodontal ligament cells.
Journal of cellular and molecular medicine (2019-06-27)
Dorottya Frank, Annamária Cser, Béla Kolarovszki, Nelli Farkas, Attila Miseta, Tamás Nagy
ZUSAMMENFASSUNG
Protein O-linked N-acetylglucosamine (O-GlcNAc) is a post-translational modification of intracellular proteins that regulates several physiological and pathophysiological process, including response to various stressors. However, O-GlcNAc's response to mechanical stress has not been investigated yet. As human periodontal ligament (PDL) cells are stimulated by compression force during orthodontic tooth movement that results in structural remodelling, in this study we investigated whether mechanical stress induces any alteration in protein O-GlcNAc in PDL cells. In this study, PDL cells isolated from premolars extracted for orthodontic indications were exposed to 0, 1.5, 3, 7 and 14 g/cm2 compression forces for 12 hours. Cell viability was measured by flow cytometry, and protein O-GlcNAc was analysed by Western blot. Cellular structure and intracellular distribution of O-GlcNAc was studied by immunofluorescence microscopy. We found that between 1.5 and 3 g/cm2 mechanical compression, O-GlcNAc significantly elevated; however, at higher forces O-GlcNAc level was not increased. We also found that intracellular localization of O-GlcNAc proteins became more centralized under 2 g/cm2 compression force. Our results suggest that structural changes stimulated by compression forces have a significant effect on the regulation of O-GlcNAc; thus, it might play a role in the mechanical stress adaptation of PDL cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone AA13, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Actin, N-terminal antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-O-Linked N-Acetylglucosamine in Maus hergestellte Antikörper, clone CTD110.6, purified from hybridoma cell culture