Merck
  • Roscovitine, a CDK5 Inhibitor, Alleviates Sevoflurane-Induced Cognitive Dysfunction via Regulation Tau/GSK3β and ERK/PPARγ/CREB Signaling.
Roscovitine, a CDK5 Inhibitor, Alleviates Sevoflurane-Induced Cognitive Dysfunction via Regulation Tau/GSK3β and ERK/PPARγ/CREB Signaling.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-11-16)
Jianhui Liu, Junjun Yang, Yinhua Xu, Gang Guo, Li Cai, Heng Wu, Yanhong Zhao, Xiaoqing Zhang
ZUSAMMENFASSUNG
Multiple exposures to anesthesia in children may increase the risk of developing cognitive impairment. Sevoflurane is an anesthetic that is commonly used in children during surgery. Cyclin-dependent kinase (CDK) 5 is involved in the regulation of sevoflurane-induced cognitive dysfunction, but the mechanistic details remain unclear. The present study evaluated the mechanism by which CDK5 mediates sevoflurane-induced cognitive dysfunction in mice. Hippocampal neurons were isolated from postnatal day 0 C57BL/6 mouse pups. Six-day-old wild-type mice were exposed to sevoflurane and then treated with the CDK5 inhibitor roscovitine. The effects on cognitive function were evaluated with the Morris water maze and neuronal damage in the hippocampus was assessed by immunohistochemical analysis. CDK5 activation increased neuronal damage by inducing Tau/glycogen synthase kinase (GSK) 3β and suppressing extracellular signal-regulated kinase (ERK)/peroxisome proliferator-activated receptor (PPAR)γ/cyclic AMP response element-binding protein (CREB) signaling following exposure to sevoflurane. CDK5 inhibition by roscovitine administration alleviated sevoflurane-induced neuronal damage and cognitive impairment. Inhibiting CDK5 with roscovitine has neuroprotective effects against neuronal injury and cognitive dysfunction caused by sevoflurane anesthesia that are exerted via modulation of Tau/GSK3β and ERK/PPARγ/CREB signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Roscovitine, ≥98% (TLC)
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder