Merck
The disintegrin domain of ADAM17 antagonises fibroblast‑carcinoma cell interactions.
International journal of oncology (2013-04-03)
Ahmad Trad, Michel Riese, Mohammad Shomali, Nina Hedeman, Timo Effenberger, Joachim Grötzinger, Inken Lorenzen
ZUSAMMENFASSUNG
The malignant phenotype of carcinoma cells depends on their ability to invade into their microenvironment promoting metastasis. Therefore, carcinoma cells overexpress many proteins, including A disintegrin and metalloproteases (ADAMs). ADAM17 is expressed by different cancer cell lines and possesses adhesive as well as enzymatic activities. To address the adhesive properties in tumour progression the recombinantly expressed soluble disintegrin domain of ADAM17 was employed. Fibroblasts and carcinoma cells adhere to the immobilized disintegrin domain. Additionally, the soluble disintegrin domain impaired fibroblast-carcinoma cell interactions and increased the shedding activity of ADAM17. Silencing of ADAM17 in fibroblasts or in carcinoma cells decreases cell-cell interaction between these cells. In summary, our results show that the adhesive properties of ADAM17 are mediated by its disintegrin domain and enables carcinoma cells to interact with their microenvironment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Integrin-β1-Antikörper, Klon P4C10, culture supernatant, clone P4C10, Chemicon®