Merck
  • Ratio of Wnt3a to BMP4 doses is critical to their synergistic effects on proliferation of differentiating mouse embryonic stem cells.
Ratio of Wnt3a to BMP4 doses is critical to their synergistic effects on proliferation of differentiating mouse embryonic stem cells.
Cell proliferation (2008-04-22)
S-Y Lin, C-L Chen, Y-L Wu, Y-C Yang, Y-M Hwu
ZUSAMMENFASSUNG
To investigate potential interactions between bone morphogenetic protein (BMP) and Wnt signalling on differentiating mouse embryonic stem cells (mESC). Mouse embryonic stem cells were cultured with differing combinations of Wnt3a, BMP4 and inhibitors of Wnt, BMP, PI-3K (phosphoinositide 3-kinase), p38, ERK1/2 (extracellular signal-regulated kinase 1/2) and JNK (c-Jun N-terminal kinase) pathways. We found that Wnt3a synergized with BMP4 to promote mESC proliferation. Furthermore, the relative ratio of Wnt3a to BMP4 doses was critical to their synergistic effects, which could be abolished by using Dkk-1, noggin or the inhibitors of PI-3K, p38, ERK1/2 and JNK pathways. We also demonstrated that combination of Wnt3a and BMP4 could suppress ectodermal differentiation of mESCs. Moreover, inhibitors of PI-3K, p38, ERK1/2 and JNK pathways could negate this effect. Relative ratio of Wnt3a to BMP4 doses is critical to their synergistic effect on differentiating mESC proliferation, which may work through PI-3K, p38, ERK1/2 and JNK pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®