- PTPN2 improves implant osseointegration in T2DM via inducing the dephosphorylation of ERK.
PTPN2 improves implant osseointegration in T2DM via inducing the dephosphorylation of ERK.
Experimental biology and medicine (Maywood, N.J.) (2019-10-17)
Ya-Nan Wang, Tingting Jia, Jiajia Zhang, Jing Lan, Dongjiao Zhang, Xin Xu
PMID31615285
MATERIALIEN