Merck
  • Upexpression of BHLHE40 in gastric epithelial cells increases CXCL12 production through interaction with p-STAT3 in Helicobacter pylori-associated gastritis.
Upexpression of BHLHE40 in gastric epithelial cells increases CXCL12 production through interaction with p-STAT3 in Helicobacter pylori-associated gastritis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
Yong-Sheng Teng, Yong-Liang Zhao, Mao-Shi Li, Yu-Gang Liu, Ping Cheng, Yi-Pin Lv, Fang-Yuan Mao, Weisan Chen, Shi-Ming Yang, Chuan-Jie Hao, Liu-Sheng Peng, Jin-Yu Zhang, Wei-Jun Zhang, Quan-Ming Zou, Yuan Zhuang
ZUSAMMENFASSUNG
BHLHE40, a member of the basic helix-loop-helix transcription factor family, has been reported to play an important role in inflammatory diseases. However, the regulation and function of BHLHE40 in Helicobacter pylori (H pylori)-associated gastritis is unknown. We observed that gastric BHLHE40 was significantly elevated in patients and mice with H pylori infection. Then, we demonstrate that H pylori-infected GECs express BHLHE40 via cagA-ERK pathway. BHLHE40 translocates to cell nucleus, and then binds to cagA protein-activated p-STAT3 (Tyr705). The complex increases chemotactic factor CXCL12 expression (production). Release of CXCL12 from GECs fosters CD4+ T cell infiltration in the gastric mucosa. Our results identify the cagA-BHLHE40-CXCL12 axis that contributes to inflammatory response in gastric mucosa during H pylori infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stra13