Merck
  • Novel Approach for Insertion of Heterologous Sequences into Full-Length ZIKV Genome Results in Superior Level of Gene Expression and Insert Stability.
Novel Approach for Insertion of Heterologous Sequences into Full-Length ZIKV Genome Results in Superior Level of Gene Expression and Insert Stability.
Viruses (2020-01-18)
Evgeniya Volkova, Konstantin A Tsetsarkin, Emilia Sippert, Felipe Assis, Guangping Liu, Maria Rios, Alexander G Pletnev
ZUSAMMENFASSUNG
Zika virus (ZIKV) emerged in the Americas in 2015, presenting unique challenges to public health. Unlike other arboviruses of the Flaviviridae family, it is transmissible by sexual contact, which facilitates the spread of the virus into new geographic areas. Additionally, ZIKV can be transmitted from mother to fetus, causing microcephaly and other severe developmental abnormalities. Reliable and easy-to-work-with clones of ZIKV expressing heterologous genes will significantly facilitate studies aimed at understanding the virus pathogenesis and tissue tropism. Here, we developed and characterized two novel approaches for expression of heterologous genes of interest in the context of full-length ZIKV genome and compared them to two previously published strategies for ZIKV-mediated gene expression. We demonstrated that among the four tested viruses expressing nLuc gene, the virus constructed using a newly developed approach of partial capsid gene duplication (PCGD) attained the highest titer in Vero cells and resulted in the highest level of nLuc expression. Suitability of the PCGD approach for expression of different genes of interest was validated by replacing nLuc sequence with that of eGFP gene. The generated constructs were further characterized in cell culture. Potential applications of ZIKV clones stably expressing heterologous genes include development of detection assays, antivirals, therapeutics, live imaging systems, and vaccines.