Merck
  • Cytochrome c'β-Met Is a Variant in the P460 Superfamily Lacking the Heme-Lysyl Cross-Link: A Peroxidase Mimic Generating a Ferryl Intermediate.
Cytochrome c'β-Met Is a Variant in the P460 Superfamily Lacking the Heme-Lysyl Cross-Link: A Peroxidase Mimic Generating a Ferryl Intermediate.
Biochemistry (2019-12-31)
Fong Ning Liew, Marisa A Brandys, Saborni Biswas, Joline N Nguyen, Mustika Rahmawati, Michael Nevala, Bradley O Elmore, Michael P Hendrich, Hyung J Kim
ZUSAMMENFASSUNG
A defining characteristic of bacterial cytochromes (cyt's) in the P460 family is an unusual cross-link connecting the heme porphyrin to the side chain of a lysyl residue in the protein backbone. Here, via proteomics of the periplasmic fraction of the ammonia-oxidizing bacterium (AOB) Nitrosomonas europaea, we report the identification of a variant member of the P460 family that contains a methionyl residue in place of the cross-linking lysine. We formally designate this protein cytochrome "c'β-Met" to distinguish it from other members bearing different residues at this position (e.g., cyt c'β-Phe from the methane-oxidizing Methylococcus capsulatus Bath). As isolated, the monoheme cyt c'β-Met is high-spin (S = 5/2). Optical spectroscopy suggests that a cross-link is absent. Hydroxylamine, the substrate for the cross-linked cyt P460 from N. europaea, did not appreciably alter the optical spectrum of cyt c'β with up to 1000-fold excess at pH 7.5. Cyt c'β-Met did however bind 1 equiv of H2O2, and with a slight excess, Mössbauer spectroscopy indicated the formation of a semistable ferryl (FeIV═O) Compound II-like species. The corresponding electron paramagnetic resonance showed a very low intensity signal indicative of a radical at g = 2.0. Furthermore, cyt c'β-Met exhibited guaiacol-dependent peroxidase activity (kcat = 20.0 ± 1.2 s-1; KM = 2.6 ± 0.4 mM). Unlike cyt c'β-Met, cyt P460 showed evidence of heme inactivation in the presence of 2 equiv of H2O2 with no appreciable guaiacol-dependent peroxidase activity. Mutagenesis of the cross-linking lysyl residue to an alanine in cyt P460, however, reversed this lack of activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
DL-Cystein, technical grade