Merck
  • Small-molecule inhibitors of ubiquitin-specific protease 7 enhance type-I interferon antiviral efficacy by destabilizing SOCS1.
Small-molecule inhibitors of ubiquitin-specific protease 7 enhance type-I interferon antiviral efficacy by destabilizing SOCS1.
Immunology (2019-11-07)
Yukang Yuan, Ying Miao, Chenhua Zeng, Jin Liu, Xiangjie Chen, Liping Qian, Xiaofang Wang, Feng Qian, Zhengyuan Yu, Jun Wang, Guanghui Qian, Qian Fu, Haitao Lv, Hui Zheng
ZUSAMMENFASSUNG
Type-I interferons (IFN-I) are used as common antiviral drugs for a range of viral diseases in clinic. However, the antiviral efficacy of IFN-I is largely restricted by negative regulators of IFN-I signaling in cells. Therefore, identification of intracellular inhibitors of IFN-I signaling is important for developing novel targets to improve IFN-I antiviral therapy. In this study, we report that the deubiquitinase ubiquitin-specific protease 7 (USP7) negatively regulates IFN-I-mediated antiviral activity. USP7 physically interacts with suppressor of cytokine signaling 1 (SOCS1) and enhances SOCS1 protein stability by deubiquitination effects, which in turn restricts IFN-I-induced activation of Janus kinase-signal transducer and activator of transcription 1 signaling. Interestingly, viral infection up-regulates USP7 and therefore facilitates viral immune evasion. Importantly, the USP7 small-molecule inhibitors P5091 and P22077 inhibit SOCS1 expression and enhance IFN-I antiviral efficacy. Our findings identify a novel regulator of IFN-I antiviral activity and reveal that USP7 inhibitors could be potential enhancement agents for improving IFN-I antiviral therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SOCS1-Antikörper, Klon 4H1, clone 4H1, Upstate®, from mouse